Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Microplate Sealing Film and Tapes
GSA_VA

Catalog No. 14222043 Shop All Axygen Products
Change view
Click to view available options
Material:
Aluminum
Litto or Ethylene or Vinyl
Polyester
Polyolefin
Rayon
Product Type:
AxySeal Sealing Film
Breathable Sealing Film
CyclerSeal Sealing Film
Heat Seal Film
Sealing Film
UltraClear Sealing Film
Dimensions (L x W):
127.2 x 79.5 mm
133.3 x 79.2 mm
135.5 x 80 mm
146 x 79.2 mm
146 x 79.6 mm
153 x 82.3 mm
Not Available
Sterility:
Non-sterile
Sterile
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Product Type Dimensions (L x W) Sterility
14-222-043 Rayon Breathable Sealing Film 153 x 82.3 mm Non-sterile
14-222-044 Rayon Breathable Sealing Film 153 x 82.3 mm Sterile
14-222-342 Aluminum Sealing Film 146 x 79.2 mm Non-sterile
14-222-343 Aluminum Sealing Film 133.3 x 79.2 mm Non-sterile
14-222-346 Polyester AxySeal Sealing Film 146 x 79.6 mm Sterile
14-222-344 Polyester AxySeal Sealing Film 146 x 79.6 mm Non-sterile
14-222-347 Polyester CyclerSeal Sealing Film 135.5 x 80 mm Non-sterile
14-222-873 Polyolefin UltraClear Sealing Film Not Available Non-sterile
14-222-191 Litto or Ethylene or Vinyl Heat Seal Film 127.2 x 79.5 mm Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-222-043 Supplier Axygen™ Supplier No. BF400
Add to Cart
Add to Cart

Offer great economy for disposable ease.

For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates. Sealing film temperature ranges from -40°C to +104°C.

Breathable

  • Allow effective gas exchange for cellular and bacterial cultivation — while preventing contamination
  • Seal polypropylene and polystyrene culture plates, 96- and 384-well plates including other assay plates
Aluminum
  • Provides exceptional seal on multi-well plates with varied temperatures and reagents
  • With compression mats, films and tapes seal during thermocycling
  • Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
  • Pierceable, PCR-AS-200
CyclerSeal
  • Exceptional seal across entire plate safeguards samples during transport or storage
  • When used with Axygen's compression mat, this polyester film eliminates well-to-well contamination and cross-over in PCR applications
AxySeal
  • Polyester-based with uniformly and consistently applied acrylic adhesive to eliminate the edge effect in sensitive ELISA assays
  • Suitable for sealing tissue culture plates, short-term storage and incubation, transport and the containment of biohazardous solutions
  • Available in both nonsterile and sterile packaging

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Material Rayon
Sterility Non-sterile
Length (English) 6.02 in.
Length (Metric) 153 mm
No. per Case 1000/Cs.
No. per Pack 100/Pk.
Dimensions (L x W) 153 x 82.3 mm
Width (English) 3.24 in.
Width (Metric) 82.3 mm
For Use With (Application) Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, allow effective gas exchange for cellular and bacterial cultivation, while preventing contamination
Product Type Breathable Sealing Film
Thickness 50 μm
Temperature Range (Metric) -20°C to +80°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.