Learn More
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
Specifications
| Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
| Color | Clear |
| Capacity (Metric) | 84 mL |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reagent Reservoir |
| No. per Case | 25/Cs. |
| No. per Pack | 25/Pk. |
| Packaging | 25/Cs. |
| Volume (Metric) Well | 22 mL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
