Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Black Hanging Format Automation Pipet Tips for Hamilton Microlab™ Star Line and Nimbus™
GREENER_CHOICE

Catalog No. 14223712 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
300 μL
1000 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Format:
Hanging Tip Rack
Stack Rack
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Filtered Format
14-223-712 1000 μL Sterile Filtered Hanging Tip Rack
14-224-026 10 μL Non-sterile Non-filtered Hanging Tip Rack
14-224-032 10 μL Non-sterile Non-filtered Stack Rack
14-224-027 10 μL Sterile Filtered Hanging Tip Rack
14-223-704 50 μL Non-sterile Non-filtered Hanging Tip Rack
14-223-706 50 μL Sterile Filtered Hanging Tip Rack
14-223-718 50 μL Non-sterile Non-filtered Stack Rack
14-223-707 300 μL Non-sterile Non-filtered Hanging Tip Rack
14-223-708 300 μL Sterile Non-filtered Hanging Tip Rack
14-223-722 300 μL Non-sterile Non-filtered Stack Rack
14-223-709 300 μL Sterile Filtered Hanging Tip Rack
14-223-711 1000 μL Sterile Non-filtered Hanging Tip Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14-223-712 Supplier Axygen™ Supplier No. HTF1000CBKHTRS
Add to Cart
Add to Cart

Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations

Axygen offers a comprehensive line of tips for Hamilton™ Microlab™ automation systems.

The tips are:

  • Manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality
  • RNase-/DNase-free and non-pyrogenic
  • Available in non-sterile, sterile, non-filtered and filtered
  • Conductive black polypropylene tips are available in 96-well hanging tip rack and nested tip rack and individual deck-seating rack format
  • Designed for Hamilton™ MICROLAB™ STAR, STARlet, STARplus and Nimbus™ workstations.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
Format Hanging Tip Rack
Tip Style Automation, Round
Packaging 1 Rack/Pk, 16 Pks/Cs.
For Use With (Equipment) Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus, Hamilton MICROLAB Nimbus
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.