Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Screw Caps with O-rings

Catalog No. 14222480 Shop All Axygen Products
Change view
Click to view available options
Color:
Blue
Brown
Clear
Green
Orange
Pink
Red
Yellow
Sterility:
Non-sterile
Sterile
Format:
Looped
Non-looped
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Format
14-222-480 Brown Non-sterile Non-looped
14-222-479 Blue Non-sterile Non-looped
14-222-484 Clear Sterile Non-looped
14-222-482 Blue Sterile Non-looped
14-222-485 Green Non-sterile Non-looped
14-222-483 Clear Non-sterile Non-looped
14-222-491 Brown Non-sterile Looped
14-222-496 Orange Non-sterile Looped
14-222-507 Red Sterile Non-looped
14-222-506 Red Non-sterile Non-looped
14-222-512 Yellow Non-sterile Non-looped
14-222-513 Yellow Sterile Non-looped
14-222-492 Clear Non-sterile Looped
14-222-497 Pink Non-sterile Looped
14-222-503 Orange Sterile Non-looped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 14-222-480 Supplier Axygen™ Supplier No. SCOBR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Select looped or non-looped format

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Material Polypropylene
Color Brown
Format Non-looped
No. per Pack 500/Pk.
No. per Case 4000/Cs.
Sterility Non-sterile
For Use With (Equipment) Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.