Learn More
Axygen™ Septa Mat for ABI multi-capillary Sequencing Instrument
Catalog No. 14222025 Shop All Axygen Products
Change view
Click to view available options
For Use With (Equipment):
ABI Multi-capillary Sequencing Instruments
Catalog No. For Use With (Equipment)
14-222-025 ABI Multi-capillary Sequencing Instruments
1 options
This item is not returnable. View return policy
Catalog No. 14-222-025 Supplier Axygen™ Supplier No. AM96SEPTA3100
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Septa Mat for ABI Multi-Capillary Sequencing Instrument
Axygen™ AxyMatsTM septa mat is designed to be used on ABI sequencer. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
  • Septa Mat for ABI Multi-Capillary Sequencing Instrument
  • For use with 96 PCR microplates
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) ABI Multi-capillary Sequencing Instruments
    Product Type Septa Mat