Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

Catalog No. 14222241 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-241 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-241 Supplier Axygen™ Supplier No. P96450VCS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For sample collection, in-vitro growth chambers and long term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.

 

  • 450 μL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SLAS 1-2004
Lid Without Lid
Array 8 x 12
Autoclavable Autoclavable
Color Clear
Product Type 96-well Storage V-bottom Microplate
Material Polypropylene
No. of Wells 96
Sterility Sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
Max. RCF 4000 x g
No. per Case 50/Cs.
No. per Pack 10/Pk.
Temperature Range (English) -112°F
Temperature Range (Metric) -80°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.