Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Storage Microplates
GREENER_CHOICE

Catalog No. 14222245 Shop All Axygen Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-245 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-245 Supplier Axygen™ Supplier No. P96450VY
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For sample collection, in-vitro growth chambers and long term storage.

Available in a variety of configurations.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SLAS 1-2004
Array 8 x 12
Autoclavable Autoclavable
Color Yellow
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
Max. RCF 4000 x g
Temperature Range (English) -112°F
Temperature Range (Metric) -80°C
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.