Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Storage Microplates
GSA_VAGREENER_CHOICE

Catalog No. 14222231 Shop All Axygen Products
Change view
Click to view available options
Color:
Blue
Clear
Sterility:
Non-sterile
Sterile
No. of Wells:
24
48
96
384
Volume (Metric) Well:
1.1 mL
1.8 mL
10,000 μL
1100 μL
125 μL
2000 μL
240 μL
5000 μL
550 μL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility No. of Wells Volume (Metric) Well
14-222-231 Clear Sterile 48 5000 μL
14-222-226 Clear Non-sterile 384 125 μL
14-222-227 Clear Sterile 384 125 μL
14-222-229 Clear Sterile 384 240 μL
14-222-228 Clear Non-sterile 384 240 μL
14-222-230 Clear Non-sterile 48 5000 μL
14-222-239 Blue Non-sterile 96 550 μL
14-222-352 Clear Sterile 96 1100 μL
14-222-350 Clear Sterile 24 10,000 μL
14-222-351 Clear Non-sterile 96 1.1 mL
14-222-349 Clear Non-sterile 24 10,000 μL
14-222-353 Clear Non-sterile 96 1.8 mL
14-222-355 Clear Sterile 96 2000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. 14-222-231 Supplier Axygen™ Supplier No. P5ML48CS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For sample collection, in-vitro growth chambers and long term storage.

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Format Standard
    Array 6 x 8
    Color Clear
    Material Polypropylene
    Volume (Metric) Well 5000 μL
    No. of Wells 48
    Sterility Sterile
    Well Shape Rectangular, Conical Bottom (V-well)
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Max. RCF 4000 x g
    No. per Case 25/Cs.
    No. per Pack 5/Pk.
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.