Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Storage Microplates
BIOPHARMAGSA_VAGREENER_CHOICE

Catalog No. 14222225 Shop All Axygen Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-225 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-225 Supplier Axygen™ Supplier No. P2MLSQCS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For sample collection, in-vitro growth chambers and long term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • 2.2 mL deep well plate
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Lid Without Lid
    Array 8 x 12
    Color Clear
    Material Polypropylene
    No. of Wells 96
    Sterility Sterile
    Well Shape Square
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    No. per Case 25/Cs.
    No. per Pack 25/Pk.
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.