Learn More
Abcam AY 9944 dihydrochloride, Hedgehog signaling inhibitor, 10MG
Supplier:  Abcam AB14588010MG
Encompass_Preferred
MW 464.3 g/mol, Purity >99%. Hedgehog signaling inhibitor. Potent Δ7-dehydrocholesterol reductase inhibitor (IC₅₀ = 13 nM). Inhibits cholesterol esterification and biosynthesis, and shows teratogenic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5044
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.