Learn More
Selleck Chemical LLC AZ 3146 S2731-1g
SDP
Supplier:  Selleck Chemical LLC S27311G
Encompass_Preferred
AZ3146 is a selective Mps1 inhibitor with IC50 of 35 nM contributes to recruitment of CENP-E (kinesin-related motor protein) less potent to FAK JNK1 JNK2 and Kit
Catalog No. 50-311-1833
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.