Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ 5704
GREENER_CHOICESDP

Catalog No. 63305
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-305 5 mg
63-302-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-305 Supplier Tocris Bioscience™ Supplier No. 6330/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective ATM kinase inhibitor; orally bioavailable

AZ 5704 is a potent and selective ATM kinase inhibitor (IC50 = 0.6 nM in an enzyme inhibition assay). Exhibits > 600-fold selectivity for ATM over other kinases. Inhibits ATM kinase in an in vitro cellular assay (IC50 = 0.33 μM). Potentiates the antitumor effects of the topoisomerase 1 inhibitor irinotecan (CPT 11, Cat. No. 2688) in tumor bearing, immunocompromised mice. Orally bioavailable.

Specifications

Chemical Name or Material 7-Fluoro-6-[6-(methoxymethyl)-3-pyridinyl]-4-[[(1S)-1-(1-methyl-1H-pyrazol-3-yl)ethyl]amino]-3-quinolinecarboxamide
CAS 1941214-06-7
Quantity 5 mg
Molecular Formula C23H23FN6O2
Formula Weight 434.47
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.