Learn More
Selleck Chemical LLC AZ 960 2mg 905586-69-8
SDP
Supplier:  Selleck Chemical LLC S22142MG
Encompass_Preferred
AZ 960 is a novel ATP competitive JAK2 inhibitor with IC50 and Ki of <3 nM and 0.45 nM, 3-fold selectivity of AZ960 for JAK2 over JAK3. AZ 960 induces apoptosis and growth arrest. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2171
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.