Learn More
Selleck Chemical LLC AZ191
SDP
Supplier:  Selleck Chemical LLC S73385MG
Encompass_Preferred
AZ191 is a potent and selective DYRK1B inhibitor with IC50 of 17 nM in a cell-free assay about 5- and 110-fold selectivity over DYRK1A and DYRK2 respectively
Catalog No. 50-217-3267
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.