Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azadibenzocyclooctyne-PEG4 biotin conjugate, Thermo Scientific™

Catalog No. AAJ65737MA
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65737MA 10 mg
AAJ65737MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65737MA Supplier Thermo Scientific Chemicals Supplier No. J65737MA

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • An ADIBO-biotin conjugate with PEG4 spacer to reduces aggregation and precipitation when labeling biological materials
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C39H51N5O8S
MDL Number MFCD22380761
Synonym ADIBO-PEG4-biotin conjugate

Specifications

Color White to Gray
Quantity 10 mg
Formula Weight 749.92
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-PEG4 biotin conjugate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.