Learn More
TARGETMOL CHEMICALS INC Azaribine 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2513025MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Azaribine (2;,3;,5;-Tri-O-acetyl-6-azauridine) is an orotate monophosphate decarboxylase (OMPD) inhibitor with broad-spectrum antiviral activity, inhibiting viral genome replication and gene transcription.Azaribine has been used in the study of Zika virus (ZIKV) infections, and in the study of psoriasis, arthritis and mycosis fungoides. Azaribine is used to study Zika virus (ZIKV) infections, psoriasis and mycosis fungoides. Purity 99.4%
Catalog No. 50-287-7532
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.