Learn More
Apexbio Technology LLC AZD-5438, 10mg. Cas: 602306-29-6 MFCD: MFCD11112135. Potent CDK1/2/9 inhibitor
SDP
Supplier:  Apexbio Technology LLC A832610
Encompass_Preferred
AZD5438 is a potent small molecule inhibitor of cyclin-dependent kinase (CDK) 1, 2 and 9 with half maximal inhibitory concentration IC50 of 16 nmol/L, 6 nmol/L and 20 nmol/L respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5111
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.