Learn More
Medchemexpress LLC HY-10422 50mg , AZD-8055 CAS:1009298-09-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1042250MG
Encompass_Preferred
HY-10422 50mg AZD-8055 CAS: 1009298-09-2 AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9168