Learn More
Selleck Chemical LLC AZD3965-5MG
SDP
Supplier:  Selleck Chemical LLC S73395MG
Encompass_Preferred
AZD3965 is a potent, selective and orally available monocarboxylate transporter 1 (MCT1) inhibitor with a binding affinity of 1.6 nM, 6-fold selective over MCT2. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1993750
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.