Learn More
Medchemexpress LLC HY-108243 5mg Medchemexpress, AZD5423 CAS:1034148-04-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1082435MG
Encompass_Preferred
Medchemexpress, HY-108243 5mg AZD5423 CAS:1034148-04-3 AZD5423 is an inhaled, potent, selective, and non-steroidal glucocorticoid receptor (GR) modulator (SGRM). AZD5423 effectively reduces allergen-induced responses in subjects with mild allergic asthma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8513
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.