Learn More
Apexbio Technology LLC AZD8055, 100mg. Cas: 1009298-09-2 MFCD: MFCD16660191. MTOR inhibitor
SDP
Supplier:  Apexbio Technology LLC A8214100
Encompass_Preferred
AZD8055 is a selective inhibitor of mTOR kinase with IC50 of 0.8 nM [1]. It competes with ATP at the ATP-binding cleft of mTOR. AZD8055 showed ~1000 fold selectivity against closely related kinases PI3K isoforms and ATM/DNA-PK [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5076
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.