Learn More
Enzo Life Sciences AZD8055 (5mg). CAS: 1009298-09-2
SDP
Supplier:  Enzo Life Sciences ENZCHM1930005
Encompass_Preferred
AZD8055 is a novel ATP-competitive inhibitor of mTOR kinase activity with an IC50 of 0.8 nM. It shows excellent selectivity (~1,000-fold) against all class I phosphatidylinositol 3-kinase (PI3K) isoforms and other members of the PI3K-like kinase family. Alternative name: CCG-168, (5-(2,4-bis((3S)-3-methylmorpholin-4-yl)pyrido(2,3-d)pyrimidin-7-yl)-2-methoxyphenyl)methanol. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (93mg/ml) or ethanol (3mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-3509
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.