Learn More
Selleck Chemical LLC Azeliragon
SDP
Supplier:  Selleck Chemical LLC S64151G
Encompass_Preferred
Azeliragon (TTP488 PF-04494700) is an orally bioavailable small molecule that inhibits the receptor for advanced glycation endproducts (RAGE) which is an immunoglobulin-like cell surface receptor overexpressed in brain tissues of patients with AD
Catalog No. 50-000-04692
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.