Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific 10% Neutral Buffered Formalin
SDP

Catalog No. 22026213
Change view
Click to view available options
Quantity:
1 gal.
2.5 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-026-213 5 gal.
22-026-354 1 gal.
22-026-350 2.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-026-213 Supplier Azer Scientific Supplier No. CUNBF5G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Available in three convenient size configurations

Specifications

Boiling Point 100°C
Quantity 5 gal.
Product Type Neutral Buffered Formalin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.