Learn More
Azer Scientific Glucose Tolerance Beverages
SDP
Catalog No. 22026204
Change view
Click to view available options
Flavor:
Fruit Punch
Lemon-Lime
Orange
Quantity:
50 g
75 g
100 g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor Quantity
22-026-204 Orange 75 g
22-026-203 Orange 50 g
22-026-205 Orange 100 g
22-026-208 Fruit Punch 100 g
22-026-207 Fruit Punch 75 g
22-026-206 Fruit Punch 50 g
22-026-209 Lemon-Lime 50 g
22-026-210 Lemon-Lime 75 g
22-026-211 Lemon-Lime 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-026-204 Supplier Azer Scientific Supplier No. 10O075
Add to Cart
Add to Cart
This item is not returnable. View return policy
Available in lemon lime, fruit punch, and orange flavors
  • Produced in an FDA certified pharmaceutical manufacturing facility
  • Dairy free, gluten free, and certified kosher product
  • Shatterproof plastic bottles
  • Manufactured according to WHO and ADA standards
  • Expiration date on product determined via rigorous accelerated and real time stability studies
  • Integrity of every batch certified by an FDA recognized independent laboratory
  • All flavors are available in standard 50, 75 and 100 gram concentrations
Specifications
Certifications/Compliance FDA, WHO, ADA
Content And Storage Room Temperature
Flavor Orange
Quantity 75 g