Learn More
Azer Scientific Glucose Tolerance Beverages
Description
- Produced in an FDA certified pharmaceutical manufacturing facility
- Dairy free, gluten free, and certified kosher product
- Shatterproof plastic bottles
- Manufactured according to WHO and ADA standards
- Expiration date on product determined via rigorous accelerated and real time stability studies
- Integrity of every batch certified by an FDA recognized independent laboratory
- All flavors are available in standard 50, 75 and 100 gram concentrations
Specifications
Specifications
| Certifications/Compliance | FDA, WHO, ADA |
| Content And Storage | Room Temperature |
| Flavor | Lemon-Lime |
| Quantity | 75 g |