Learn More
Sigma Organic Chemistry Azetidine | 1G | 503-29-7 | MFCD00005165
Supplier: Sigma Organic Chemistry 2810691G

CAS #: 503-29-7
MDL #: MFCD00005165
Purity: >98 %
UNSPSC Code: 12352100
Molecular Weight: 57.09
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.