Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azilsartan 98.0+%, TCI America™
SDP

Catalog No. A26011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26011G 1 g
A2601200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26011G Supplier TCI America Supplier No. A26011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 147403-03-0
Molecular Formula C25H20N4O5
Molecular Weight (g/mol) 456.46
MDL Number MFCD20278186
InChI Key KGSXMPPBFPAXLY-UHFFFAOYSA-N
Synonym azilsartan, unii-f9nux55p23, azilsartan tak-536, chembl57242, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzimidazole-7-carboxylic acid, 1h-benzimidazole-7-carboxylic acid, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy, 1-2'-2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl 1,1'-biphenyl-4-yl methyl-2-ethoxy-1h-benzimidazole-7-carboxylic acid, 2-ethoxy-1-2'-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl-1h-benzo d imidazole-7-carboxylic acid, 2-ethoxy-1-2'-4,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl biphenyl-4-yl methyl benzimidazole-7-carboxylic acid, 2-ethoxy-3-4-2-5-oxo-2h-1,2,4-oxadiazol-3-yl phenyl phenyl methyl benzimidazole-4-carboxylic acid
PubChem CID 9825285
ChEBI CHEBI:68850
IUPAC Name 2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid
SMILES CCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1

Specifications

Melting Point 188°C
Color White
Quantity 1 g
Formula Weight 456.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Azilsartan
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.