Learn More
Apexbio Technology LLC Azlocillin sodium salt 37091-65-9 1g
Supplier: Apexbio Technology LLC B18931000
Azlocillin sodium salt (CAS 37091-65-9) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby impeding cross-linking of peptidoglycan layers and inducing bacterial lysis Azlocillin sodium salt exerts its biological activity primarily through inhibition of PBPs disrupting cell wall integrity in bacteria In microbiological assays azlocillin demonstrates broad-spectrum antibacterial activity against both Gram-positive and Gram-negative bacteria reported IC50 values vary by microbial species and experimental conditions typically determined in minimum inhibitory concentration (MIC) assays Based on these pharmacological properties azlocillin sodium salt holds research potential in microbiological and biomedical investigations including studies of bacterial susceptibility resistance mechanisms and combinational antimicrobial therapies
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.