Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Azlon™ Polypropylene, Autoclavable Carboy
SCI_ED

Catalog No. S01989E Shop All Dynalab Corporation Products
Change view
Click to view available options
Capacity (Metric):
5 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01989E 20 L
S01989C 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S01989E Supplier Dynalon™ Supplier No. 5056346
Add to Cart
Add to Cart

For Use With: Storing and dispensing liquids in educational settings.; Autoclavable; Closure Material: Polypropylene; Color: White

  • Autoclavable polypropylene, heavy-duty construction
  • Stylish rounded octagonal shape for increased rigidity when full
  • Offset neck for easier filling and pouring
  • Integral, comfortable blow-molded handles for ease of carrying; handles are end-sealed to prevent contamination
  • Heavy-duty closures
  • Molded graduations allow easy viewing of contents

 

Specifications

Autoclavable Autoclavable
Capacity (English) 5.3 gal.
Capacity (Metric) 20 L
Closure Material Polypropylene
Color White
For Use With (Application) Storing and dispensing liquids in educational settings.
Includes Blow molded handles
Material Polypropylene
Neck Style Offset
Product Type Carboy
Shape Rounded Octagonal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.