Learn More
Azobenzene, 97+%, Thermo Scientific Chemicals
Description
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Azobenzene is used to construct photoswitch tethered ligands to turn protein activity on and off. It is also used to increase the extracellular or intracellular concentration of active molecules to study the mechanism of action of sympathetic and parasympathetic neuromediators.
Solubility
Soluble in alcohol, ether, benzene and glacial acetic acid. Insoluble in water.
Notes
Store at 4°C. Incompatible with strong oxidizing agents. Air and light sensitive.
Specifications
Melting Point | 65°C to 69°C |
Density | 1.203 |
Boiling Point | 293°C |
Quantity | 100 g |
UN Number | UN3077 |
Merck Index | 14,917 |
Solubility Information | Soluble in alcohol,ether,benzene and glacial acetic acid. Insoluble in water. |
Formula Weight | 182.23 |
Percent Purity | ≥97% |
Chemical Name or Material | Azobenzene |
Safety and Handling
Hazard Category | H302+H332-H340-H373 |
Hazard Statement | GHS H Statement H350-H341-H373-H302-H332 May cause cancer. Suspected of causing genetic defects. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 203-102-5 |
RTECSNumber | CN1400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.