Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azobenzene 98.0+%, TCI America™
SDP

Catalog No. A0565500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0565500G 500 g
A056525G 25 g
A0565100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0565500G Supplier TCI America Supplier No. A0565500G

Chemical Identifiers

CAS 103-33-3
Molecular Formula C12H10N2
Molecular Weight (g/mol) 182.23
MDL Number MFCD00003022
InChI Key DMLAVOWQYNRWNQ-UHFFFAOYSA-N
Synonym azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
PubChem CID 2272
ChEBI CHEBI:190358
IUPAC Name diphenyldiazene
SMILES C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 68°C
Boiling Point 293°C
Quantity 500 g
UN Number 3077
Formula Weight 182.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene
EINECSNumber (3)-0361
RTECSNumber CN1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.