Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azobisisobutyronitrile

Catalog No. 768375100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-837-5100ML 100 mL
76-837-5500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-837-5100ML Supplier Sigma Aldrich Supplier No. 768375100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Azobisisobutyronitrile (AIBN) is an azo-compound and is widely used as a free radical initiator. This compound has labile carbon-nitrogen covalent bond which undergoes homolytic scission under thermal, chemical or photochemical conditions producing free radicals. They are useful in many reactions like halogenation, polymerisation of vinyl monomers, grafting reactions, curing of rubbers and unsaturated polymers and cross-linking of polyolefins.Used as an initiator in the synthesis of highly cross-linked Poly(divinylbenzene) (PDVB) polymers. Used as an initiator in the polymerization process of 2-hydroxyethyl methacrylate (HEMA).

Specifications

CAS 78-67-1
Density 0.808 g/mL (at 25°C)
Molecular Formula C8H12N4
Refractive Index n20/D 1.368
Linear Formula C8H12N4
Quantity 100 mL
MDL Number MFCD00013808
Synonym 2,2 -Azobis(2-methylpropionitrile) solution; radical initiator; alpha,alpha,-Azoisobutyronitrile solution; AIBN solution
Molecular Weight (g/mol) 164.21
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.