Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azomethine H, MP Biomedicals™

Catalog No. ICN15416905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15416905 5 g
ICN15416901 1 g
ICN15416910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15416905 Supplier MP Biomedicals, Inc Supplier No. 0215416905
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5941-07-1
Molecular Formula C17H14NNaO9S2
Molecular Weight (g/mol) 463.41
MDL Number MFCD00149588
InChI Key BEDIAJJRAYEWBI-TTWKNDKESA-M
Synonym 8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate

Specifications

Assay Percent Range 1.04
Extinction Coefficient Extinction Coefficient max: 237nm C=0.01% ≥50, 000 (Lit.)
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.[Na].[Na]
IUPAC Name 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium
PubChem CID 123134089
Formula Weight 423.4
Color Orange
Quantity 5 g
Physical Form Powder
Chemical Name or Material Azomethine H, Chemistry
EINECSNumber 227-698-1
Recommended Storage Store at 4°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.