Learn More
ATCC Lacticaseibacillus rhamnosus
Supplier:  ATCC 7469A
Encompass
Lacticaseibacillus rhamnosus was derived from an existing strain. This bacterium can be used in assays of folic acid and it produces easily dispersed clumps when grown in (BBL) folic-acid assay medium.
Catalog No. 50-238-0017
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.