Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aztreonam, 95.7%, MP Biomedicals™

Catalog No. ICN15041501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15041501 1 g
ICN15041505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15041501 Supplier MP Biomedicals, Inc Supplier No. 0215041501
Add to Cart
Add to Cart

Specifically targets penicillin-binding protein 3 (PBP-3) of Gram-negative bacteria

  • Displays high degree of resistance to β-lactamase degradation
  • With high specificity for gram-negative aerobic rods
  • White Powder
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
Synonym Azthreonam, Azactam
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Type Aztreonam
Physical Form Crystalline Powder
Color White
Quantity 1 g
Assay Percent Range 0.957
Formula Weight 435.4
Chemical Name or Material Aztreonam
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.