Learn More
Cayman Chemical ParItaprevIr 25mg
SDP
Supplier:  Cayman Chemical 2830125 MG
Encompass_Preferred
An acaricide has broad-spectrum activity against various plant-feeding mite species lethal to wild-type but not chs1 mutant T urticae mite eggs (LC50s 1 2 and 5000 mg/L respectively) induces formation of thyroid tumors in male rats from 13-1000 mg/kg per day
Catalog No. 50-293-8192
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.