Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azulene, 99%

Catalog No. AAL0827103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0827103 1 g
AAL08271MD 250 mg
AAL0827106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0827103 Supplier Thermo Scientific Chemicals Supplier No. L0827103
Add to Cart

CAS: 275-51-4 | C10H8 | 128.174 g/mol

Azulene is used as an additive in ointments. It is an active ingredient in cosmetics, shaving creams and other topicals. It is used as antioxidant and in anti-inflammatory activities. It is involved in the colorimetric determination of furfural. It acts as ligand for low valent metals and forms pi complexes with both cyclopentadienyl and cylcloheptatrienyl ligands in organometallic chemistry. It is also used as precursor for the preparation of (azulene)Mo2 (CO)6 and (azulene)Fe2 (CO)5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azulene is used as an additive in ointments. It is an active ingredient in cosmetics, shaving creams and other topicals. It is used as antioxidant and in anti-inflammatory activities. It is involved in the colorimetric determination of furfural. It acts as ligand for low valent metals and forms pi complexes with both cyclopentadienyl and cylcloheptatrienyl ligands in organometallic chemistry. It is also used as precursor for the preparation of (azulene)Mo2 (CO)6 and (azulene)Fe2 (CO)5.

Solubility
Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 275-51-4
Molecular Formula C10H8
Molecular Weight (g/mol) 128.174
MDL Number MFCD00003810
InChI Key CUFNKYGDVFVPHO-UHFFFAOYSA-N
Synonym cyclopentacycloheptene, azunamic, bicyclo 5.3.0 decapentaene, azulen, unii-82r6m9mglp, bicyclo 5.3.0-1,3,5,7,9-decapentaene, bicyclo 5.3.0-deca-2,4,6,8,10-pentaene, 82r6m9mglp, azulekeep
PubChem CID 9231
ChEBI CHEBI:31249
IUPAC Name azulene
SMILES C1=CC=C2C=CC=C2C=C1

Specifications

Melting Point 98°C to 101°C
Boiling Point 241°C to 243°C
Quantity 1 g
Beilstein 969517
Merck Index 14,926
Solubility Information Soluble in organic solvents.
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material Azulene
Hazard Category H315-H320-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-993-6
RTECSNumber CO4570000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.