Learn More
Azure Biosystems AzureCyto In-Cell Western Kit - GAM 700, GAR 800
SDP
Catalog No. AC2023AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
AC2023AZ Each
1 options
Catalog No. AC2023AZ Supplier Azure Biosystems Supplier No. AC2023
Add to Cart
Add to Cart
Kit with AzureSpectra 700 Goat anti-Mouse and AzureSpectra 800 Goat anti-Rabbit. Provides all reagents necessary to perform 5 96-well In-Cell Western Assays with total cell staining.
  • Kit with AzureSpectra 700 Goat anti-Mouse and AzureSpectra 800 Goat anti-Rabbit. Provides all reagents necessary to perform 5 96-well In-Cell Western Assays with total cell staining.
  • Includes AzureCyto Permeabilization Solution, AzureCyto Block Solution, AzureCyto Total Cell Stain, AzureSpectra 700 Goat anti-Mouse and AzureSpectra 800 Goat anti-Rabbit secondary antibodies.
  • Reagents to perform up to five 96-well assays.
Specifications
Product Type Western Blot Kit
For Use With (Application) Western Blot
Quantity Each