Learn More
Azure Biosystems AzureSpot Pro 20
SDP
Catalog No. AC1132AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Product Type: Software
Catalog No.
AC1132AZ Product Type: Software
1 options
Catalog No. AC1132AZ Supplier Azure Biosystems Supplier No. AC1132
Analysis of 1D gels and Western blots, band analysis, array analysis, colony counting.
  • Analysis of 1D gels and Western blots, band analysis, array analysis, colony counting.
  • PC and Mac compatible.
  • Electronic License with 20 computer installations.
Specifications
Product Type Software