Learn More
Azure Biosystems AzureSpot Pro
SDP
Catalog No. AC1078AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Product Type:
Software
Catalog No. Product Type
AC1078AZ Software
1 options
Catalog No. AC1078AZ Supplier Azure Biosystems Supplier No. AC1078
Analysis of 1D gels and Western blots, band analysis, array analysis, colony counting.
  • Analysis of 1D gels and Western blots, band analysis, array analysis, colony counting.
  • PC and Mac compatible.
  • Electronic License with 4 computer installations.
Specifications
Product Type Software