Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Azurocidin Monoclonal Antibody (5F1)
GREENER_CHOICE

Catalog No. MA571721
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA571721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA571721 Supplier Invitrogen™ Supplier No. MA571721
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Azurocidin Monoclonal Antibody for ELISA, IHC (F), IHC (P), Western Blot

Azurocidin, also called heparin-binding protein (HBP), is an inactive homolog of serine proteinases residing in granulocytes. Initially it was thought to have only activity against gram-negative bacteria. Later it was also found to act against gram-positive bacteria and C. albicans. Azurocidin is considered a family member of polymorphonuclear leukocytes- derived antimicrobial proteins like defensins, LL-37 and lysozyme. Azurocidin has been recognized as a player in the activation and modulation of the immune response and may act to alarm the immune system. The cationic domain at one side of the protein is essential for its antimicrobial activity. It is stored in azurophil granules as well as secretory granules and as a result partly released at early stage of extravasation. Azurocidin is involved in chemotaxis and activation of monocytes, cytokine release and phagocytosis leading to more efficient bacterial clearance. The primary targeted environment of azurocidin are cells in the bloodstream, that is the endothelial lining, and the extravascular surroundings. The interaction of azurocidin with leucocytes is mediated via beta 2-intergrins. High plasma levels of azurocidin might help with the identification of patients which are at risk in developing sepsis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Azurocidin
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Antibody with no preservative, pH 7.4
Gene AZU1
Gene Accession No. P20160
Gene Alias AZAMP; AZU; AZU1; Azurocidin; azurocidin 1; CAP37; cationic antimicrobial protein 37; cationic antimicrobial protein CAP37; HBP; Heparin-binding protein; hHBP; HUMAZUR; NAZC; neutrophil azurocidin; pHBP
Gene Symbols AZU1
Host Species Mouse
Immunogen Recombinant Human Azurocidin Protein
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 566
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.