Learn More
MARSHALL SCIENTIFIC Buchi B-490 Heating Bath
SDP
Supplier:  MARSHALL SCIENTIFIC BUB490
Encompass
The Buchi B-490 Heating Bath can accommodate evaporating flasks ranging in size from 50 to 3000mL, with an overall holding capacity of 5L. This heating bath can be adjusted from 20° to 180°C and has a temperature accuracy of + 1ºC, making it suitable for high/low boiling solvents. The Buchi B-490’s aluminum heating pan features a corrosion-resistant PTFE coating and, for added safety, an insulated outer wall and two overtemperature shutoffs. The waterbath features a digital LED display, which shows real-time temperature and setpoint. (The B-490 is for use with and included in evaporating systems R-200 and R-205.)
Catalog No. NC2591263
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.