Learn More
United Scientific™ B.O.D. Bottles, Numbered
SDPSCI_ED
Catalog No. p-200009368 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
300 mL
No. Sequence:
1 to 24
25 to 48
49 to 72
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) No. Sequence
S15755 300 mL 1 to 24
S012435 300 mL 25 to 48
S012434 300 mL 49 to 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S15755 Supplier United Scientific™ Supplier No. BOD300N
Only null left
Add to Cart
Add to Cart
Efficiently conduct the 5-day biochemical oxygen demand (BOD) test with United Scientific™ BOD Bottles, designed for incubating diluted samples of clean, waste, effluent, and polluted water. Feature a barcode for streamlined tracking and glass stoppers for secure sealing.
  • Bottles are numbered to allow for convenient organization of samples
  • 300 mL bottles also include a barcode
  • Glass robotic stoppers are included
  • 300 mL line is available in a series of numbers from 1 through 120 in increments of 24
  • Made of borosilicate glass
  • Easy-to-read numbers
  • Barcode for easy recording
  • Write on marking spaces
Specifications
Volume (Metric) 300 mL
No. Sequence 1 to 24
Stopper Glass Robotic