Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam B-Phycoerythrin Conjugation Kit, Lightning-Link, 3 x 60 μg
Description
- Labeling time: < 4 hrs
- Hands-on time: 30 secs
- Antibody recovery: 100%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Antibody |
| Shipping Condition | Ambient |
| Label or Dye | B-Phycoerythrin |
| Product Type | Protein Labeling Reagent |
| Alternate Part Number | ab102871 |
| Content And Storage | Store at -20°C. |
| Format | Kit |
| Quantity | 3 x 60 μg |
| For Use With (Application) | Antibody conjugation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?