Learn More
Abcam B-Phycoerythrin Conjugation Kit, Lightning-Link™, 60 μg
Description
- Labeling time: < 4 hrs
- Hands-on time: 30 secs
- Antibody recovery: 100%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Target Molecule | Antibody |
| Shipping Condition | Ambient |
| Product Type | Protein Labeling Reagent |
| Format | Kit |
| Label or Dye | B-Phycoerythrin |
| Detection Method | Fluorescence |
| Quantity | 60 μg |
| For Use With (Application) | Antibody labeling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.