Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam B-Phycoerythrin Conjugation Kit, Lightning-Link, 600 μg

Catalog No. 502585609
Encompass_Preferred
Change view
Click to view available options
Quantity:
600 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-5609 600 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-5609 Supplier Abcam Supplier No. AB102871600UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid antibody labeling in under 4 hours with 100% recovery and minimal hands-on time.

  • Labeling time: < 4 hrs
  • Hands-on time: 30 secs
  • Antibody recovery: 100%
  • Storage temperature: -20°C
  • Shipping temperature: Ambient (Cannot Ship with Ice)

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Antibodies
Shipping Condition Ambient
Label or Dye B-Phycoerythrin
Product Type Protein Labeling Reagent
Content And Storage Store at -20°C.
Format Kit
Detection Method Fluorescence
Quantity 600 μg
For Use With (Application) Antibody conjugation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.