Learn More
Description
ToughBond™ Ultra Aggressive Polyester (B-483) is perfect for labeling products and general ID and can be applied to powder coated, smooth, textured or irregular surfaces. A ribbon is required to print this material.
- Glossy white polyester with permanent rubber-based adhesive is thermal transfer printable
- Material is designed for high adhesion to textured metals and low surface energy plastics
- UL approved, CSA accepted
- Wide service temperature range
Specifications
Specifications
| Adhesive Type | Permanent (Rubber-based) |
| Color | Red on White |
| Finish | Gloss |
| Format | Die-cut |
| Material | Polyester (B-483) |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Sterility | Non-sterile |
| Display Text | DANGER |
| For Use With (Application) | Barcode Labeling, Equipment Labeling, Data and Telecommunications Labeling, Inventory and Inspection Labeling, General Identification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.