Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GALNT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733702 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3702 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3702 Supplier Proteintech Group Inc Supplier No. 220481AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

B3GALNT1 gene ontology annotations related to this gene include protein glycosylation.

Specifications

Antigen B3GALNT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene B3GALNT1
Gene Accession No. O75752, Q6AY39, Q920V1
Gene Alias b3Gal T3, B3GALNT1, B3GALT3, Beta 1,3 GalNAc T1, Beta 1,3 GalTase 3, Beta 3 Gx T3, beta3Gal T3, Beta3GalT3, galT3, Gb4Cer, GLCT3, GLOB, Globoside synthase, P, P1
Gene Symbols B3GALNT1
Host Species Rabbit
Immunogen B3GALNT1 Fusion Protein Ag16976
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26879, 310508, 8706
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.