Learn More
Invitrogen™ B3GNT4 Polyclonal Antibody

Description
Immunogen sequence: MLPPQPSAAH QGRGGRSGLL PKGPAMLCRL CWLVSYSLAV LLLGCLLFLR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 92%; Rat: 86%.
Specifications
Specifications
| Antigen | B3GNT4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | B3GNT4 |
| Gene Accession No. | Q9C0J1 |
| Gene Alias | 1010001G17Rik; B3GNT4; B3GN-T4; beta-1,3-Gn-T4; beta-1,3-N-acetylglucosaminyltransferase 4; beta-1,3-N-acetylglucosaminyltransferase bGn-T4; beta3Gn-T4; BGnT-4; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 4; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4; UNQ1898/PRO4344 |
| Gene Symbols | B3GNT4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.